Colchicine 97,0 (contains 5% Ethyl Acetate at maximum)

Supplier: TCI America
Danger

Synonyms: 7α-H-Colchicine, (S)-(-)-Colchicine

C0380-500MG C0380-5G
TCC0380-500MGEA 53 USD
TCC0380-500MG TCC0380-5G
Colchicine 97,0 (contains 5% Ethyl Acetate at maximum)
Colchicine

CAS Number: 64-86-8 MDL Number: MFCD00078484 Molecular Formula: C22H25NO6 Molecular Weight: 399.44 Purity/Analysis Method: 97.0% (HPLC) Form: Crystal Melting point (°C): 150 Specific rotation [a]20/D: -238 deg (C=1, EtOH)

Formula: C₂₂H₂₅NO₆
MW: 399.44 g/mol
Melting Pt: 150 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078484
CAS Number: 64-86-8
EINECS: 200-598-5
UN: 1544
ADR: 6.1 I
Merck Index: 12,02536
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fe9c85ba71f6cdab2382a1d99d970b4be7a6f31&vid=2fe9c85ba71f6cdab2382a1d99d970b4be7a6f31&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR