Choline chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-hydroxy-N,N,N-trimethylethanaminium chloride

C0329-25G C0329-500G
TCC0329-025GEA 29.06 USD
TCC0329-025G TCC0329-500G
Choline chloride ≥98.0% (by titrimetric analysis)
Choline chloride

CAS Number: 67-48-1
MDL Number: MFCD00011721
Molecular Formula: C5H14ClNO
Molecular Weight: 139.62
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 302


Product Categories: Bioscience, Other Plant Hormones, Vitamin Related Compounds
Other Product Names / Synonyms: (2-Hydroxyethyl)trimethylammonium Chloride

Formula: HOH₂CCH₂N(CH₃)₃Cl
MW: 139.62 g/mol
Melting Pt: 302 °C
Density: ∼1.205 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011721
CAS Number: 67-48-1
EINECS: 200-655-4
Merck Index: 13,02226
https://us.vwr.com/stibo/web/std.lang.all/73/69/28937369.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR