4-Chloropyridine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 4-Chloropyridinium chloride

C0281-25G C0281-250G
TCC0281-025GEA 41.8 USD
TCC0281-025G TCC0281-250G
4-Chloropyridine hydrochloride ≥98.0%
4-Chloropyridine hydrochloride

CAS Number: 7379-35-3 MDL Number: MFCD00012829 Molecular Formula: C5H4ClN Molecular Weight: 150.00 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal

Formula: C₅H₅Cl₂N
MDL Number: MFCD00012829
CAS Number: 7379-35-3
EINECS: 230-946-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c315c6ffcea118609f377b883e0039af118863b&vid=8c315c6ffcea118609f377b883e0039af118863b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR