Chlorocyclohexane ≥98.0%

Supplier: TCI America

Synonyms: Cyclohexyl chloride

C0153-500ML C0153-25ML
TCC0153-500MLEA 69.73 USD
TCC0153-500ML TCC0153-25ML
Chlorocyclohexane ≥98.0%
Chlorocyclohexane

CAS Number: 542-18-7 MDL Number: MFCD00003822 Molecular Formula: C6H11Cl Molecular Weight: 118.60 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 143 Melting point (°C): -44 Flash Point (°C): 32 Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: Cyclohexyl Chloride

Formula: C₆H₁₁Cl
Boiling Pt: 143 °C
Melting Pt: –44 °C
Density: 1.00 (20 °C)
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00003822
CAS Number: 542-18-7
EINECS: 208-806-6
UN: 1993
ADR: 3,III
Merck Index: 13,02761
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9209010a5de1ec60eecb19ca9eefdda05fa6eba1&vid=9209010a5de1ec60eecb19ca9eefdda05fa6eba1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR