4-Chlorobenzonitrile ≥98.0%

Supplier: TCI America
Warning

Synonyms: p-Chlorobenzonitrile

C0135-25G C0135-500G
TCC0135-025GEA 23 USD
TCC0135-025G TCC0135-500G
4-Chlorobenzonitrile ≥98.0%
4-Chlorobenzonitrile

CAS Number: 623-03-0 MDL Number: MFCD00001813 Molecular Formula: C7H4ClN Molecular Weight: 137.57 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 223 Melting point (°C): 92 Flash Point (°C): 108

Formula: C₇H₄ClN
MW: 137.57 g/mol
Boiling Pt: 223
Melting Pt: 92 °C
Density: 1.1133 g/cm³ (16.8 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00001813
CAS Number: 623-03-0
EINECS: 210-765-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a25a1ffead86cc1df1b1404140ace516ec8a0bb3&vid=3e7fca588c19d807812aff92d7fbd4762a952e2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR