(1S)-(-)-Borneol ≥95.0%
Supplier: TCI America
Synonyms:
(1S-endo)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
B1012-25G
B1012-500G
TCB1012-500GEA
296.29
USD
TCB1012-500G
TCB1012-25G
(1S)-(-)-Borneol ≥95.0%
(1S)-(-)-Borneol
CAS Number: 464-45-9
MDL Number: MFCD00003759
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 207
Flash Point (°C): 65
Specific rotation [a]20/D: -36 deg (C=5, EtOH)
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Boiling Pt: 203 °C (1013 hPa) Melting Pt: 207 °C Density: 1.011 g/cm³ (20 °C) Flash Pt: 65 °C |
MDL Number:
MFCD00003759 CAS Number: 464-45-9 EINECS: 207-353-1 UN: 1312 ADR: 4.1,III Merck Index: 13,01328 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...