Bismarck brown Y

Supplier: TCI America

Synonyms: 1,3-Bis(2,3-diaminophenylazo)benzene dihydrochloride 4,4'-[1,3-Phenylenebis(azo)]bisbenzene-1,3-diamine dihydrochloride

B0780-25G
TCB0780-025GEA 31 USD
TCB0780-025G
Bismarck brown Y
Bismarck Brown Y

CAS Number: 10114-58-6
MDL Number: MFCD00012977
Molecular Formula: C18H18N8
Molecular Weight: 419.31
Form: Crystal
Color: Deep Yellow Red


Other Product Names / Synonyms: Basic Brown 1, Vesuvine G

Formula: C₁₈H₂₀Cl₂N₈
MW: 419.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012977
CAS Number: 10114-58-6
EINECS: 233-314-3
Merck Index: 12,01294
https://us.vwr.com/stibo/web/std.lang.all/27/01/28932701.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR