Butyric acid ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Butanoic acid, n-Butyric acid
B0754-500ML
B0754-25ML
TCB0754-500MLEA
28.96
USD
TCB0754-500ML
TCB0754-25ML
Butyric acid ≥99.0%
Butyric acid
CAS Number: 107-92-6
MDL Number: MFCD00002814
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method:
Formula:
C₄H₈O₂ MW: 88.11 g/mol Boiling Pt: 163.5 Melting Pt: -3 Density: 0.96 (20 °C) Flash Pt: 75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002814 CAS Number: 107-92-6 EINECS: 203-532-3 UN: 2820 ADR: 8 III Merck Index: 13,01593 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...