tert-Butanol ≥99.0%

Supplier: TCI America
Danger

Synonyms: Trimethylcarbinol, tert-Butyl alcohol, 2-Methyl-2-propanol

B0706-25ML B0706-500ML
TCB0706-25MLEA 31.81 USD
TCB0706-25ML TCB0706-500ML
tert-Butanol ≥99.0%
tert-Butanol

CAS Number: 75-65-0 MDL Number: MFCD00004464 Molecular Formula: C4H10O Molecular Weight: 74.12 Purity/Analysis Method: 99.0% (GC) Form: Crystal Boiling point (°C): 82 Flash Point (°C): 11 Freezing point (°C): 25


Product Categories: Bioscience, Treatment Agents (Staining) Other Product Names / Synonyms: tert-Butanol, 2-Methyl-2-propanol

Formula: (CH₃)₃COH
MW: 74.12 g/mol
Boiling Pt: 82.2
Melting Pt: 25.3
Density: 0.7887 g/cm³ (20 °C)
Flash Pt: 11
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS Number: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3 II
Merck Index: 12,01577
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28f5aa2b3070472cbde2f95a3591455aeea52615&vid=28f5aa2b3070472cbde2f95a3591455aeea52615&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR