(±)-2-Butanol ≥99.0%

Supplier: TCI America
Danger

Synonyms: (±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol

B0705-25ML B0705-500ML
TCB0705-25MLEA 20.35 USD
TCB0705-25ML TCB0705-500ML
(±)-2-Butanol ≥99.0%
(±)-2-Butanol

CAS Number: 78-92-2 MDL Number: MFCD00004569 Molecular Formula: C4H10O Molecular Weight: 74.12 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 100 Melting point (°C): -115 Flash Point (°C): 24 Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: sec-Butyl Alcohol

Formula: H₃CCH₂CH(OH)CH₃
MW: 74.12 g/mol
Boiling Pt: 94
Melting Pt: -115
Density: 0.81 (20 °C)
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00004569
CAS Number: 78-92-2
EINECS: 201-158-5
UN: 1120
ADR: 3 III
Merck Index: 13,01540
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47347320002d3593ecb0b9319c821663510fdfdd&vid=b16f9f7c5badc0f8bd3cdf5b7175d907e0133f7c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR