Bromotrichloromethane ≥98.0%

Supplier: TCI America
Warning

B0662-500G B0662-25G
TCB0662-500GEA 115 USD
TCB0662-500G TCB0662-25G
Bromotrichloromethane ≥98.0%
Bromotrichloromethane

CAS Number: 75-62-7 MDL Number: MFCD00000783 Molecular Formula: CBrCl3 Molecular Weight: 198.27 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 104 Melting point (°C): -6 Specific Gravity (20/20): 2.02

Formula: BrCl₃C
Boiling Pt: 104 °C
Melting Pt: –6 °C
Density: 2.02 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000783
CAS Number: 75-62-7
EINECS: 200-886-0
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bef6e36eaf3a0a15f1fa40a266911ce19e5e6ac5&vid=25499f34c9cc42d8624ed6f4d727fde1f7b0d216&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR