1-Bromopentane ≥98.0%
Supplier: TCI America
|
Danger
|
B0628-500ML
B0628-25ML
TCB0628-500MLEA
86.46
USD
TCB0628-500ML
TCB0628-25ML
1-Bromopentane ≥98.0%
1-Bromopentane
CAS Number: 110-53-2
MDL Number: MFCD00000267
Molecular Formula: C5H11Br
Molecular Weight: 151.05
Purity/Analysis Method:
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Amyl Bromide, Pentyl Bromide
Formula:
BrH₂C(CH₂)₃CH₃ MW: 151.05 g/mol Boiling Pt: 129 Melting Pt: -95 Density: 1.22 (20 °C) Flash Pt: 31 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000267 CAS Number: 110-53-2 EINECS: 203-776-0 UN: 1993 ADR: 3 III Merck Index: 13,00606 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...