Bromoethane ≥99.0%

Supplier: TCI America
Danger

Synonyms: Bromic ether, Monobromoethane, Ethyl bromide, Hydrobromic ether

B0588-500G B0588-25G
TCB0588-500GEA 51.67 USD
TCB0588-500G TCB0588-25G
Bromoethane ≥99.0%
Bromoethane

CAS Number: 74-96-4 MDL Number: MFCD00000232 Molecular Formula: C2H5Br Molecular Weight: 108.97 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 38 Flash Point (°C): -23 Specific Gravity (20/20): 1.46 Storage Temperature: 0-10°C


Other Product Names / Synonyms: Ethyl Bromide

Formula: BrH₂CCH₃
Boiling Pt: 38 °C
Melting Pt: –118 °C
Density: 1.46 (20 °C)
Flash Pt: –23 °C
Storage Temperature: Ambient
MDL Number: MFCD00000232
CAS Number: 74-96-4
EINECS: 200-825-8
UN: 1891
ADR: 6.1,II
Merck Index: 13,03807
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab1b352213c0bde14a9dbfe44c14950bf6fa339d&vid=795be899d8fd7d1436754b2b1fdfad61241b2171&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR