Disulfiram ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B0479-25G B0479-500G
TCB0479-025GEA 19 USD
TCB0479-025G TCB0479-500G
Disulfiram ≥97.0% (by titrimetric analysis)
Disulphiram

CAS Number: 97-77-8 MDL Number: MFCD00009048 Molecular Formula: C10H20N2S4 Molecular Weight: 296.52 Purity/Analysis Method: 97.0% (T) Form: Crystal Melting point (°C): 70


Product Categories: Synthetic Organic Chemistry, Thiocarbonyl Compounds, Materials Chemistry, Precise Polymerization Reagents, Bioscience, Inhibitors (for Pharmacological Research), Other Pharmaceutical Ingredients Other Product Names / Synonyms: Bis(diethylthiocarbamoyl) Disulfide, Disulfiram

Formula: C₁₀H₂₀N₂S₄
Boiling Pt: 369 °C (1013 hPa)
Melting Pt: 70 °C
Density: 1.34 g/cm³ (20 °C)
MDL Number: MFCD00009048
CAS Number: 97-77-8
EINECS: 202-607-8
UN: 3077
ADR: 9,III
Merck Index: 13,03399
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b58b436e004291aceefd763b0cd9a00f5b908ac&vid=8b58b436e004291aceefd763b0cd9a00f5b908ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR