Biphenyl (PCB No. 0) ≥99.5%

Supplier: TCI America
Warning

B0465-500G B0465-25G
TCB0465-500GEA 43.18 USD
TCB0465-500G TCB0465-25G
Biphenyl (PCB No. 0) ≥99.5%
Biphenyl (PCB No. 0)

CAS Number: 92-52-4 MDL Number: MFCD00003054 Molecular Formula: C12H10 Molecular Weight: 154.21 Purity/Analysis Method: 99.5% (GC) Form: Crystalline Powder Boiling point (°C): 256 Melting point (°C): 70 Flash Point (°C): 113

Formula: C₁₂H₁₀
Boiling Pt: 256 °C
Melting Pt: 70 °C
Density: 1.175 g/cm³ (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00003054
CAS Number: 92-52-4
EINECS: 202-163-5
UN: 3077
ADR: 9,III
Merck Index: 13,03346
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=417ad70b9bfe414891502b3b2a8630510216507f&vid=417ad70b9bfe414891502b3b2a8630510216507f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR