Benzalkonium chloride 50% in water
Supplier: TCI America
|
Danger
|
B0414-500G
B0414-25G
TCB0414-500GEA
117.87
USD
TCB0414-500G
TCB0414-25G
Benzalkonium chloride 50% in water
Benzalkonium chloride
CAS Number: 63449-41-2
MDL Number: MFCD00050250
Form: Clear Liquid
Color: Colorless
Other Product Names / Synonyms: Alkyldimethylbenzylammonium Chloride, Benzyldimethylalkylammonium Chloride
Formula:
C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇) Storage Temperature: Ambient |
MDL Number:
MFCD00145757 CAS Number: 63449-41-2 EINECS: 264-151-6 UN: 2923 ADR: 8,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...