1-Bromotetradecane ≥97.0%

Supplier: TCI America

Synonyms: Myristyl bromide

B0233-25ML B0233-500ML
TCB0233-500MLEA 93.97 USD
TCB0233-500ML TCB0233-025ML
1-Bromotetradecane ≥97.0%
1-Bromotetradecane

CAS Number: 112-71-0
MDL Number: MFCD00000228
Molecular Formula: C14H29Br
Molecular Weight: 277.29
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 158
Freezing point (°C): 5
Specific Gravity (20/20): 1.02


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Myristyl Bromide, Tetradecyl Bromide

Formula: BrH₂C(CH₂)₁₂CH₃
MW: 277.29 g/mol
Boiling Pt: 205 °C
Melting Pt: 5 °C
Density: 1.02 (20 °C)
Flash Pt: 158 °C
Storage Temperature: Ambient
MDL Number: MFCD00000228
CAS Number: 112-71-0
EINECS: 203-999-3
https://us.vwr.com/stibo/web/std.lang.all/23/09/28932309.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR