Ammonium acetate ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Acetic acid ammonium salt

A2269-250G
TCA2269-250GEA 26.91 USD
TCA2269-250G
Ammonium acetate ≥97.0% (by titrimetric analysis)
Ammonium acetate

CAS Number: 631-61-8
MDL Number: MFCD00013066
Molecular Formula: C2H4O2
Molecular Weight: 77.08
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 114

Formula: H₃CCOONH₄
MW: 77.08 g/mol
Melting Pt: 114 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://us.vwr.com/stibo/web/std.lang.all/04/45/28930445.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR