2,2,2-Trifluoroethyl acrylate ≥98.0% stabilized
Supplier: TCI America
Synonyms:
Acrylic acid 2,2,2-trifluoroethyl ester
Stabilization:
Stabilised with Stabilised
(stabilized with MEHQ)
CAS Number: 407-47-6
MDL Number: MFCD00000444
Molecular Formula: C5H5F3O2
Molecular Weight: 154.09
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): 13
Specific Gravity (20/20): 1.23
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid 2,2,2-Trifluoroethyl Ester
Formula:
C₅H₅F₃O₂ MW: 154.09 g/mol Boiling Pt: 92 °C Density: 1.23 (20 °C) Flash Pt: 13 °C |
MDL Number:
MFCD00000444 CAS Number: 407-47-6 EINECS: 206-987-6 UN: 3272 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...