4-Amino-2,2,6,6-tetramethylpiperidine ≥98.0%

Supplier: TCI America

A1077-25ML A1077-500ML
TCA1077-25MLEA 59.6 USD
TCA1077-25ML TCA1077-500ML
4-Amino-2,2,6,6-tetramethylpiperidine ≥98.0%
4-Amino-2,2,6,6-tetramethylpiperidine

CAS Number: 36768-62-4 MDL Number: MFCD00005984 Molecular Formula: C9H20N2 Molecular Weight: 156.27 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 93 Flash Point (°C): 81 Freezing point (°C): 5 Specific Gravity (20/20): 0.90

Formula: C₉H₂₀N₂
MW: 156.27 g/mol
Boiling Pt: 93 °C
Density: 0.90 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00005984
CAS Number: 36768-62-4
EINECS: 253-197-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96aa30713e106d036c97f9fda97ac2329d151c15&vid=96aa30713e106d036c97f9fda97ac2329d151c15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR