Alginic acid

Supplier: TCI America

A0733-25G A0733-500G
TCA0733-025GEA 30 USD
TCA0733-025G TCA0733-500G
Alginic acid
Alginic acid

CAS Number: 9005-32-7 MDL Number: MFCD00081309 Form: Crystal Color: White


Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Natural Polymers (for DDS)

Formula: (C₆H₈O₆)n
Storage Temperature: Ambient
MDL Number: MFCD00081309
CAS Number: 9005-32-7
EINECS: 232-680-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR