L(+)-Asparagine monohydrate ≥99.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(S)-(+)-Asparagine monohydrate
A0542-250G
A0542-25G
TCA0542-250GEA
420.21
USD
TCA0542-250G
TCA0542-025G
L(+)-Asparagine monohydrate ≥99.0% (by HPLC, titration analysis)
L(+)-Asparagine monohydrate
CAS Number: 5794-13-8
MDL Number: MFCD00151038
Molecular Formula: C4H8N2O3
Molecular Weight: 132.12
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 235
Specific rotation [a]20/D: 31 deg (C=10, HCl(1+3))
Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Asn-OH
Formula:
C₄H₈N₂O₃·1H₂O MW: 150.13 g/mol Melting Pt: 235 °C Density: 1.567 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00151038 CAS Number: 5794-13-8 EINECS: 200-735-9 Merck Index: 13,00842 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...