3-Aminophenol ≥98.5% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
m-Aminophenol
, 3-Hydroxyaniline
A0383-25G
A0383-500G
TCA0383-25GEA
57.44
USD
TCA0383-25G
TCA0383-500G
3-Aminophenol ≥98.5% (by GC, titration analysis)
3-Aminophenol
CAS Number: 591-27-5
MDL Number: MFCD00007786
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 164
Melting point (°C): 122
Flash Point (°C): 165
Formula:
C₆H₇NO MW: 109.13 g/mol Boiling Pt: 164 °C Melting Pt: 122 °C Density: 1.275 g/cm³ (20 °C) Flash Pt: 165 °C |
MDL Number:
MFCD00007786 CAS Number: 591-27-5 EINECS: 209-711-2 UN: 2512 ADR: 6.1,III Merck Index: 13,00459 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...