6-Aminocaproic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
6-Aminohexanoic acid
A0312-500G
A0312-25G
TCA0312-025GEA
33.5
USD
TCA0312-025G
TCA0312-500G
6-Aminocaproic acid ≥98.0% (by titrimetric analysis)
6-Aminocaproic acid
CAS Number: 60-32-2
MDL Number: MFCD00008238
Molecular Formula: C6H13NO2
Molecular Weight: 131.18
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 204
Formula:
C₆H₁₃NO₂ MW: 131.17 g/mol Boiling Pt: 257 °C (1013 hPa) Melting Pt: 204 °C Flash Pt: 207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008238 CAS Number: 60-32-2 EINECS: 200-469-3 Merck Index: 13,00431 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...