Aminoacetaldehyde diethyl acetal ≥97.0%

Supplier: TCI America

Synonyms: 2,2-Diethoxyethanamine, 2-Aminoacetaldehyde diethylacetal

A0247-250ML A0247-25ML
TCA0247-250MLEA 368 USD
TCA0247-250ML TCA0247-25ML
Aminoacetaldehyde diethyl acetal ≥97.0%
Aminoacetaldehyde diethyl acetal

CAS Number: 645-36-3 MDL Number: MFCD00008136 Molecular Formula: C6H15NO2 Molecular Weight: 133.19 Purity/Analysis Method: 97.0% (GC,T) Form: Clear Liquid Boiling point (°C): 163 Flash Point (°C): 45 Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: Aminoacetaldehyde Diethyl Acetal

Formula: C₆H₁₅NO₂
Boiling Pt: 163 °C
Density: 0.92 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00008136
CAS Number: 645-36-3
EINECS: 211-439-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17c318b565848dcf91a2bb48e84c73872365765d&vid=17c318b565848dcf91a2bb48e84c73872365765d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR