Adiponitrile ≥98.0%
Supplier: TCI America
Synonyms:
Hexanedinitrile
A0168-500ML
A0168-25ML
TCA0168-500MLEA
52
USD
TCA0168-500ML
TCA0168-25ML
Adiponitrile ≥98.0%
Adiponitrile
CAS Number: 111-69-3
MDL Number: MFCD00001975
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method:
Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: 1,4-Dicyanobutane, Hexanedinitrile, Tetramethylene Dicyanide
Formula:
C₆H₈N₂ Boiling Pt: 295 °C Melting Pt: 2 °C Density: 0.96 (20 °C) Flash Pt: 93 °C |
MDL Number:
MFCD00001975 CAS Number: 111-69-3 EINECS: 203-896-3 UN: 2205 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...