Acetylcholine chloride ≥98.0%

Supplier: TCI America

Synonyms: (2-Acetoxyethyl)trimethylammonium chloride, 2-(Acetyloxy)-N,N,N-trimethylethanaminium chloride, 2-Acetoxy-N,N,N-trimethylethanaminium chloride

A0084-25G A0084-500G
TCA0084-025GEA 48 USD
TCA0084-025G TCA0084-500G
Acetylcholine chloride ≥98.0%
Acetylcholine chloride

CAS Number: 60-31-1 MDL Number: MFCD00011698 Molecular Formula: C7H16ClNO2 Molecular Weight: 181.66 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 150

Formula: H₃CC(O)O(CH₂)₂N(CH₃)₃Cl
Melting Pt: 150 °C
Density: 1.263 g/cm³ (20 °C)
MDL Number: MFCD00011698
CAS Number: 60-31-1
EINECS: 200-468-8
Merck Index: 13,00089
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bc3437a14cfb351bab83f6706546b6583a0ccb8&vid=7bc3437a14cfb351bab83f6706546b6583a0ccb8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR