Roof Attachment, Ace Glass
Supplier: Ace Glass
7508-06 7508-08
89059-250EA 502.99 USD
89059-250 89059-252
Roof Attachment, Ace Glass
Fittings and Connectors
Roof attachment is designed for securing a sampling cane to the roof of a van
It is fabricated from nylon, with two O-Rings to ensure a watertight seal. Threaded length is 20.3 cm (8").