Magnetic Spinbar, Round, Ace Glass Incorporated
Supplier: Ace Glass
13664-01 13664-02 13664-03 13664-04
89084-198EA 35.34 USD
89084-198 89084-204 89084-210 89084-218
Magnetic Spinbar, Round, Ace Glass Incorporated
Stirrers Magnetic Stirring Bars
Borosilicate glass-coated magnetic stirring bars are great for use in high temperatures, (up to 275°C) or for applications where zero absorption of the stirred solution is needed.
Order Now
SPECIFICATIONS
Environmentally Preferable