Sodium iodide ACS, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: IodurilHydriodic acid sodium saltSodium monoiodide

BDHVBDH9294-500G
BDH9294-500GCS 5029.87 USD
BDH9294-500G
Sodium iodide ACS, VWR Chemicals BDH®
Sodium iodide

White solid; may become brown on exposure to air

Formula: NaI
MW: 149.89 g/mol
Boiling Pt: 1300 °C (1013 hPa)
Melting Pt: 662 °C
Density: 3.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9,III
Merck Index: 13,08705

Order Now

Specification Test Results

Barium = 0.002 %
Calcium = 0.002 %
Chloride & Bromide = 0.01 %
Heavy Metals (as Pb) = 5 ppm
Insolubles = 0.01 %
Iodate = 3 ppm
Iron = 5 ppm
Magnesium = 0.001 %
pH (5%, Water) @25C 6.0 - 9.0
Phosphate = 0.001 %
Potassium = 0.01 %
Purity = 99.5 %
Sulfate = 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR