5-Bromo-2'-deoxyuridine (broxuridine) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Bromo-2'-deoxyuridine
, 5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione
H27260-06
H27260-03
H27260-MD
AAH27260-03EA
125.43
USD
AAH27260-03
AAH27260-MD
AAH27260-06
5-Bromo-2'-deoxyuridine (broxuridine) 99%
5-Bromo-2'-deoxyuridine (Broxuridine)
Formula:
C₉H₁₁BrN₂O₅ MW: 307.10 g/mol Melting Pt: ca 190 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006529 CAS Number: 59-14-3 EINECS: 200-415-9 |
Specification Test Results
Optical rotation | +31° (c=1 in 0.1N HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...