Hydrogen chloride 1 M in diethyl ether

Supplier: Thermo Scientific Chemicals
Danger

H27125-AP H27125-AE
AAH27125-AEEA 41.63 USD
AAH27125-AE AAH27125-AP
Hydrogen chloride 1 M in diethyl ether
Hydrogen chloride
Formula: HCl
MW: 36.46 g/mol
Density: 0.731
Flash Pt: -40 °C(-40 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 2924
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR