N,N-Diethylhydroxylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

B23302-14 B23302-22
AAB23302-14EA 37.3 USD
AAB23302-14 AAB23302-22
N,N-Diethylhydroxylamine 97%
N,N-Diethylhydroxylamine

Fieser: 6,187 19,127


Purity: 97% CAS No.: 3710-84-7 Molecular Formula: C4H11NO Formula Weight: 89.14 Flash Point: 46°(114°F) Refractive Index: 1.4215 Melting Point: -25° Boiling Point: 125-130° Beilstein Reference: 1731349 UN No.: 1993 MDL No.: MFCD00002126

Formula: C₄H₁₁NO
Boiling Pt: 125…130 °C
Melting Pt: –25 °C
Density: 0.867
Flash Pt: 46 °C(114 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002126
CAS Number: 3710-84-7
EINECS: 223-055-4
UN: 1993
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR