Barium hydroxide monohydrate ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
045888-22
045888-36
AA45888-22EA
28.6
USD
AA45888-22
AA45888-36
Barium hydroxide monohydrate ≥95%
Barium hydroxide monohydrate
Purity: 95%
CAS No.: 22326-55-2
Molecular Formula: H2Ba2O2.H2O
Formula Weight: 189.38 (171.36anhy)
Physical Form: Crystalline
Density: 3.743
Appearance: White
Odor: Odorless
UN No.: 3262
MDL No.: MFCD00149151
Formula:
H₂BaO₂·1H₂O Density: 3.743 Storage Temperature: Ambient |
MDL Number:
MFCD00149151 CAS Number: 22326-55-2 EINECS: 241-234-5 UN: 3262 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...