Sulfuric acid ≥72% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

045596-K2 045596-K7
AA45596-K2EA 159 USD
AA45596-K2 AA45596-K7
Sulfuric acid ≥72% (w/w) in aqueous solution
Sulfuric acid

CAS No.: 7664-93-9 Molecular Formula: H2O4S Formula Weight: 98.08 Physical Form: Liquid Appearance: Colorless to pale yellow Odor: Acrid UN No.: 1830 MDL No.: MFCD00064589

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR