Devarda's alloy (Cu:Al:Zn 50:45:5 wt%), granular

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Copper aluminium zinc alloy (50:45:5 w%)Devarda's metal

045526-22 045526-36
AA45526-22EA 53.37 USD
AA45526-22 AA45526-36
Devarda's alloy (Cu:Al:Zn 50:45:5 wt%), granular
Devarda's alloy
Boiling Pt: 906 °C (1013 hPa)
Melting Pt: 490…560 °C
Density: 4.355
Storage Temperature: Ambient
MDL Number: MFCD00198176
CAS Number: 8049-11-4
UN: 3089
ADR: 4.1,II
Merck Index: 14,2941

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR