Silver ≥99.9% (metals basis), nanopowder APS 20-40 nm
Supplier: Thermo Scientific Chemicals
045509-14
045509-06
AA45509-14EA
662
USD
AA45509-14
AA45509-06
Silver ≥99.9% (metals basis), nanopowder APS 20-40 nm
Silver
Purity: 99.9%
CAS No.: 7440-22-4
Molecular Formula: Ag
Physical Form: No Oxide layer
Odor: Characteristic
UN No.: 3077
MDL No.: MFCD00003397
Formula:
Ag Storage Temperature: Ambient |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 EINECS: 232-033-3 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...