Iron ≥99.9% (metals basis), nanopowder APS 10-30 nm

Supplier: Thermo Scientific Chemicals
Danger

045507-06 045507-14
AA45507-06EA 305.44 USD
AA45507-06 AA45507-14
Iron ≥99.9% (metals basis), nanopowder APS 10-30 nm
Iron

Purity: 99.9% CAS No.: 7439-89-6 Molecular Formula: Fe(core)/FeO(shell) Physical Form: FeO layer 0.5-1.5nm Sensitivity: Air Sensitive Appearance: Grey UN No.: 3089 MDL No.: MFCD00010999

Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
UN: 3089
ADR: 4.1 II
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR