Copper ≥99.9% (metals basis), nanopowder APS 20-50 nm

Supplier: Thermo Scientific Chemicals
Danger

045504-06 045504-14
AA45504-06EA 237 USD
AA45504-06 AA45504-14
Copper ≥99.9% (metals basis), nanopowder APS 20-50 nm
Copper

Purity: 99.9% CAS No.: 7440-50-8 Molecular Formula: Cu(core)/CuO(shell) Physical Form: CuO layer approx. 2nm Appearance: Black Odor: Odorless UN No.: 3089 MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 II
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR