Tungsten ≥99% (metals basis), powder -50+100 mesh

Supplier: Thermo Scientific Chemicals
Warning

045379-22 045379-36 045379-A3
AA45379-22EA 62.6 USD
AA45379-22 AA45379-36 AA45379-A3
Tungsten ≥99% (metals basis), powder -50+100 mesh
Tungsten

Purity: 99+% CAS No.: 7440-33-7 UN No.: 3089 MDL No.: MFCD00011461

Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
UN: 3089
ADR: 4.1 III
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR