Holmium, Plasma Standard Solution, Specpure®, Ho 10 µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
045255-AE
AA45255-AEEA 217 USD
AA45255-AE
Holmium, Plasma Standard Solution, Specpure®, Ho 10 µg/ml, Thermo Scientific
Standards Cationic Standards Holmium Standards
Molecular Formula: Ho2O3 in 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011049
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR