Glassy carbon, rods, Diameter 6 mm (0.24 in) Type 1

Supplier: Thermo Scientific Chemicals

045004-DJ 045004-DM
AA45004-DJEA 282 USD
AA45004-DJ AA45004-DM AA45004-DR
Glassy carbon, rods, Diameter 6 mm (0.24 in) Type 1
Glassy carbon

Physical Form: ≈0.44g/cm MDL No.: MFCD00133992

Formula: C
Melting Pt: 3652 °C
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR