Aluminum, AAS standard solution, Specpure®, Al 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
033557-AE
033557-AP
AA33557-AEEA
44.43
USD
AA33557-AE
AA33557-AP
Aluminum, AAS standard solution, Specpure®, Al 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Aluminum Standards
Molecular Formula: Al in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00134029
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...