Benzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
030749-A5
030749-30
030749-A1
AA30749-A1EA
52
USD
AA30749-A1
AA30749-30
AA30749-A5
Benzaldehyde 98%
Benzaldehyde
Formula:
C₆H₅CHO MW: 106.12 g/mol Boiling Pt: 178…179 °C Melting Pt: –26 °C Density: 1.047 Flash Pt: 62 °C (143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003299 CAS Number: 100-52-7 EINECS: 202-860-4 UN: 1990 ADR: 9,III Merck Index: 13,01057 |
![]() |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...