Mercury, plasma standard solution, Specpure®, Hg 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
013865-AD
013865-AE
013865-AP
AA13865-ADEA
52.9
USD
AA13865-AD
AA13865-AE
AA13865-AP
Mercury, plasma standard solution, Specpure®, Hg 1000µg/ml, Thermo Scientific
Standards
Cationic Standards
Mercury Standards
Molecular Formula: Hg in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011035
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...