Samarium, Plasma Standard Solution, Specpure®, Sm 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
13854-AP 013854-AE 013854-AD 13854-AD 13854-AE 13854-AP
AA13854-AEEA 76.9 USD
AA13854-AE AA13854-AD CAAA13854-AE CAAA13854-AD AA13854-AP CAAA13854-AP
Samarium, Plasma Standard Solution, Specpure®, Sm 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Samarium Standards
Molecular Formula: Sm2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011233
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR