2-Methoxy-3-pyridineboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20094-06
L20094-03
AAAL20094-06EA
258
USD
AAAL20094-06
AAAL20094-03
2-Methoxy-3-pyridineboronic acid 98%
2-Methoxy-3-pyridineboronic acid
Purity: 98%
CAS No.: 163105-90-6
Molecular Formula: C6H8BNO3
Formula Weight: 152.95
Melting Point: 136-138°
MDL No.: MFCD03411572
Formula:
C₆H₈BNO₃ Melting Pt: 136…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411572 CAS Number: 163105-90-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...