Ultramark® 1621, Ultramark® mass spec standard

Supplier: Thermo Scientific Chemicals
Warning

L16698-03 L16698-MD
AAAL16698-03EA 141.01 USD
AAAL16698-03 AAAL16698-MD
Ultramark® 1621, Ultramark® mass spec standard
Ultramark® 1621

CAS No.: 105809-15-2 Formula Weight: 2120.00 Refractive Index: 1.3530 MDL No.: MFCD00242454

Formula: C₆H₁₈N₃P₃
Density: 1.8
Storage Temperature: Ambient
MDL Number: MFCD00242454
CAS Number: 105809-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR