Hexafluorophosphoric acid ∼60% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

L15728-AE L15728-AC L15728-AP
AAAL15728-AEEA 68.45 USD
AAAL15728-AE AAAL15728-AC AAAL15728-AP
Hexafluorophosphoric acid ∼60% (w/w) in aqueous solution
Hexafluorophosphoric acid

Fieser: 1,42 2,402


CAS No.: 16940-81-1 Molecular Formula: HPF6 Formula Weight: 145.97 UN No.: 1782 MDL No.: MFCD00011350

Formula: HF₆P
Density: 1.65
Storage Temperature: Ambient
MDL Number: MFCD00011350
CAS Number: 16940-81-1
EINECS: 241-006-5
UN: 1790
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR