Boranepyridine complex 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Pyridine Borane, Borane, pyridine complex
L13178-09
L13178-18
AAAL13178-09EA
57.6
USD
AAAL13178-09
AAAL13178-18
Boranepyridine complex 95%
Boranepyridine complex
Fieser: 8,50 9,59 11,69 12,65 18,58 19,43 21,65
Purity: 95%
CAS No.: 110-51-0
Molecular Formula: C5H8BN
Formula Weight: 92.93
Storage Temperature: 2°C to 8°C
Flash Point: 73°(163°F)
Refractive Index: 1.5320
Odor: Unpleasant
Melting Point: 9-11°
Boiling Point: 65°/1mm
Beilstein Reference: 3674913
UN No.: 2810
MDL No.: MFCD00012435
Formula:
C₅H₈BN MW: 92.94 g/mol Boiling Pt: 65 °C (1 mmHg) Melting Pt: 9…11 °C Density: 0.920 Flash Pt: 73 °C (163 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00012435 CAS Number: 110-51-0 EINECS: 203-773-4 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...